Methyl-1-methylcyclopropyl ketone 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Methyl 1-methylcyclopropyl ketone, 1-(1-Methylcyclopropyl)ethanone

147630050 147630250
ACRO147630050EA 68.8 EUR
ACRO147630050 ACRO147630250
Methyl-1-methylcyclopropyl ketone 95%
Methyl-1-methylcyclopropyl ketone

Appearance: Clear colorless to yellow Liquid

Formel: C₆H₁₀O
Molecular Weight: 98.145 g/mol
Siedepunkt: 125…128 °C (760 mmHg)
Dichte: 0.895 g/cm³
Flammpunkt: 24 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010245
CAS-Nummer: 1567-75-5
UN: 1224
ADR: 3 III

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4320 to 1.4350 (20°C, 589 nm)
GC ≥94 %

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR