1-(Adamantan-1-yl)ethanon 99%

Lieferant: Thermo Scientific

Synonyme: 1-Adamantyl Methyl Ketone, Methyl 1-Adamantyl Ketone

164740010 164740050 164740250
ACRO164740010EA 24.3 EUR
ACRO164740010 ACRO164740050 ACRO164740250
1-(Adamantan-1-yl)ethanon 99%
1-Acetyladamantane

Appearance: White Crystalline powder

Formel: C₁₂H₁₈O
Molecular Weight: 178,28 g/mol
Schmelzpunkt: 53…55 °C
Flammpunkt: 108 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00074739
CAS-Nummer: 1660-04-4

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 53 to 55 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR