N-Ethylcarbazole-3-carboxaldehyde 94% tech.
Lieferant: Thermo Scientific
Synonyme:
3-Formyl-N-ethylcarbazole
166030250
166031000
166035000
ACRO166030250EA
102
EUR
ACRO166030250
ACRO166031000
ACRO166035000
N-Ethylcarbazole-3-carboxaldehyde 94% tech.
N-Ethylcarbazole-3-carboxaldehyde
CAS No.: 7570-45-8
Formel:
C₁₅H₁₃NO Molecular Weight: 223,28 g/mol Schmelzpunkt: 85…91 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00004963 CAS-Nummer: 7570-45-8 |
Spezifikation Testergebnisse
Appearance | Yellow to ochre Crystalline powder or granules |
Infrared spectrum | Conforms |
Melting point | 85 to 91 °C |
HPLC | ≥92 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...