(E)-4-Phenylbut-3-enoic acid 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: trans-Styrylacetic acid, trans-4-Phenylbut-3-enoic acid, trans-4-Phenyl-3-butenoic acid

168100100 168100010
ACRO168100100EA 184 EUR
ACRO168100100 ACRO168100010
(E)-4-Phenylbut-3-enoic acid 96%
(E)-4-Phenylbut-3-enoic acid

CAS No.: 1914-58-5

Formel: C₁₀H₁₀O₂
Molecular Weight: 162,188 g/mol
Schmelzpunkt: 82…88 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00002783
CAS-Nummer: 1914-58-5

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow-brown Crystalline powder or needles
Infrared spectrum Conforms
Melting point 82 to 88 °C
HPLC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR