Potassium oxonate 97.5%

Lieferant: Thermo Fisher Scientific

Synonyme: Potassium Allantoxanate, Oxonic acid Potassium Salt, Potassium 4,6-Dihydroxy-1,3,5-triazine-2-carboxylate, Allantoxanic acid Potassium Salt, Potassium 4,6-dioxo-1,4,5,6-tetrahydro-1,3,5-triazine-2-carboxylate

168530250 168531000
ACRO168530250EA 109 EUR
ACRO168530250 ACRO168531000
Potassium oxonate 97.5%
Potassium oxonate

Appearance: White Crystalline powder

Formel: C₄H₃N₃O₄
Molecular Weight: 195.18 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010565
CAS-Nummer: 2207-75-2

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥96.50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR