(S)-2-Amino-6-guanidinohexanoic acid hydrochloride 98%
Lieferant: Thermo Scientific
Synonyme:
H-HoArg-OH·HCl, L(+)-Homoarginine hydrochloride, H-HomoArg-OH.HCl, H-HomoArg-OH·HCl
169090010
169090050
ACRO169090010EA
65.1
EUR
ACRO169090010
ACRO169090050
(S)-2-Amino-6-guanidinohexanoic acid hydrochloride 98%
(S)-2-Amino-6-guanidinohexanoic acid hydrochloride
CAS No.: 1483-01-8
Formel:
C₇H₁₆N₄O₂·HCl Molecular Weight: 224,7 g/mol Schmelzpunkt: 213…215 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012622 CAS-Nummer: 1483-01-8 |
Spezifikation Testergebnisse
Appearance | White Granular powder |
Infrared spectrum | Conforms |
Water | ≤2 % (K.F.) |
Specific optical rotation | +17° to +19° (20°C, 589 nm) (c=1, 1 N HCl) |
Titration with HClO4 | ≥97.5 % (On dried substance) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...