Methyl-(triphenylphosphoranyliden)acetat 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Methyl (triphenylphosphoranylidene)acetate, Methyl 2-(triphenylphosphoranylidene)acetate, Methoxycarbonylmethylene-Triphenylphosphorane
169560250
169561000
169565000
ACRO169560250EA
105
EUR
ACRO169560250
ACRO169561000
ACRO169565000
Methyl-(triphenylphosphoranyliden)acetat 98%
Methyl-2-(triphenylphosphoranylidene)acetate
CAS No.: 2605-67-6
Formel:
C₂₁H₁₉O₂P Molecular Weight: 334,36 g/mol Schmelzpunkt: 164…169 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00008455 CAS-Nummer: 2605-67-6 UN: 2811 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Melting point | 164 to 169 °C |
HPLC | ≤2 % triphenylphosphine + triphenylphosphine oxide |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...