1-Phenylcyclopropanecarbonitrile 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1-Phenylcyclopropyl cyanide, 1-Phenyl-1-cyclopropanecarbonitrile, 1-Cyano-1-phenylcyclopropane

170590100 170590500
ACRO170590100EA 35.3 EUR
ACRO170590100 ACRO170590500
1-Phenylcyclopropanecarbonitrile 97%
1-Phenylcyclopropanecarbonitrile

Appearance: Clear colorless to light yellow Liquid

Formel: C₁₀H₉N
Molecular Weight: 143,189 g/mol
Siedepunkt: 133…137 °C (30 mmHg)
Dichte: 1 g/cm³
Flammpunkt: >110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001270
CAS-Nummer: 935-44-4
UN: 2810
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5370 to 1.5400 (20°C, 589 nm)
GC ≥96.0 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR