Sulphobromophthalein sodium hydrate
Lieferant: Thermo Scientific
|
Gefahr
|
173400050
173400250
ACRO173400050EA
290
EUR
ACRO173400050
ACRO173400250
Sulphobromophthalein sodium hydrate
Sulphobromophthalein sodium hydrate
Appearance: White to off-white to grey Powder
Formel:
C₂₀H₈Br₄Na₂O₁₀S₂·xH₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD16556312 CAS-Nummer: 123359-42-2 |
Spezifikation Testergebnisse
Appearance | White to off-white to grey Powder |
Infrared spectrum | Conforms |
Water | ≤8 % (K.F.) |
HPLC | ≥95 % |
UV Lambda max | 575 to 581 nm (c=10 mg/l, 0.1N NaOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...