DL-2-Allylglycin 99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
DL-2-Amino-4-pentenoic acid, DL-2-Allylglycine, 2-Amino-4-pentenoic Acid, 2-Aminopent-4-enoic acid
174220010
174220050
ACRO174220010EA
67.4
EUR
ACRO174220010
ACRO174220050
DL-2-Allylglycin 99%
DL-2-Allylglycine
CAS No.: 7685-44-1
Formel:
C₅H₉NO₂ Molecular Weight: 115,13 g/mol Schmelzpunkt: 251…253 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00063103 CAS-Nummer: 7685-44-1 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥98.5 % |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...