5-Methyl-5-phenylhydantoin 99%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Methyl-5-phenylimidazolidine-2,4-dione

177680050
ACRO177680050EA 40.1 EUR
ACRO177680050
5-Methyl-5-phenylhydantoin 99%
5-Methyl-5-phenylhydantoin

CAS No.: 6843-49-8

Formel: C₁₀H₁₀N₂O₂
Molecular Weight: 190,202 g/mol
Schmelzpunkt: 199…201 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005265
CAS-Nummer: 6843-49-8

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 198 to 201 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR