PTFE

Lieferant: Thermo Scientific

Synonyme: Polytetrafluoroethylene, Teflon

178930050 178931000 178932500
ACRO178930050EA 22 EUR
ACRO178930050 ACRO178931000 ACRO178932500
PTFE
PTFE

CAS No.: 9002-84-0

Formel: (CF₂CF₂)n
Siedepunkt: >400 °C (760 mmHg)
Schmelzpunkt: 327 °C
Dichte: 2 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00084455
CAS-Nummer: 9002-84-0

Bestellen

Spezifikation Testergebnisse

Appearance White Powder
Infrared spectrum Conforms
Loss on drying ≤1 %
HPLC ≥97.5 %
Residue after ign. (sulfated) ≤0.25 %
Specific optical rotation -33° to -37° (20°C, 589 nm) (1 % in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR