4-O-Methyldopamine hydrochloride 98%
Lieferant: Thermo Scientific
|
Achtung
|
187740500
187745000
ACRO187740500EA
81.4
EUR
ACRO187740500
ACRO187745000
4-O-Methyldopamine hydrochloride 98%
4-O-Methyldopamine hydrochloride
CAS No.: 645-33-0
Formel:
C₉H₁₄ClNO₂ Schmelzpunkt: 207…211 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012896 CAS-Nummer: 645-33-0 |
Spezifikation Testergebnisse
Appearance | White to Light beige Powder |
Infrared spectrum | Conforms |
Melting point | 207 to 211 °C |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...