Rose Bengal 85%, rein, hohe Reinheit biologische Färbemittel

Lieferant: Thermo Scientific
Achtung

Synonyme: Sodium 4,5,6,7-tetrachloro-2',4',5',7'-tetraiodo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate), Food Red No. 105, CI 45440, Acid red 94, Aniline Rose, Rose Bengal sodium

189450010 189450050 189450250 189451000 189455000
ACRO189450010EA 25.5 EUR
ACRO189450010 ACRO189450050 ACRO189450250 ACRO189451000 ACRO189455000
Rose Bengal 85%, rein, hohe Reinheit biologische Färbemittel
Rose Bengal

CAS No.: 632-69-9

Formel: C₂₀H₂Cl₄I₄O₅Na₂
Molecular Weight: 1017,63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00005043
CAS-Nummer: 632-69-9

Bestellen

Spezifikation Testergebnisse

Appearance Bordeaux to red-brown Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤10 % (105°C)
Dye content ≥80 %
UV Lambda max 540 to 550 nm (in water)
Absorption Ratio P-15/P+15 1.21 to 1.95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR