Sodium-3-methyl-2-oxobutanoate 98+%

Lieferant: Thermo Scientific

Synonyme: 3-Methyl-2-oxobutanoic acid, sodium salt, Sodium 3-methyl-2-oxobutanoate

189720050 189720250 189720010
ACRO189720050EA 214 EUR
ACRO189720050 ACRO189720250 ACRO189720010
Sodium-3-methyl-2-oxobutanoate 98+%
Sodium-3-methyl-2-oxobutanoate

CAS No.: 3715-29-5

Formel: C₅H₇NaO₃
Molecular Weight: 138,098 g/mol
Schmelzpunkt: 227…231 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002581
CAS-Nummer: 3715-29-5

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 227 to 231 °C
Titration with HClO4 ≥98.0 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR