Chromeazurol S (Mordant blue 29)
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2,6-Dichlorodimethyl-sulphoxy-fuchsonedicarboxylic acid sodium salt, 5-((3-Carboxy-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)(2,6-dichloro-3-sulphophenyl)methyl)-2-hydroxy-3-methylbenzoic acid,trisodium salt, Mordant Blue 29, 5-((3-Carboxy-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)(2,6-dichloro-3-sulphophenyl)methyl)-2-hydroxy-3-methylbenzoic acid, trisodium salt, Chrome Azurol S
190050250
190051000
ACRO190050250EA
96.8
EUR
ACRO190050250
ACRO190051000
Chromeazurol S (Mordant blue 29)
Chromeazurol S (Mordant blue 29)
CAS No.: 1667-99-8
Formel:
C₂₃H₁₆Cl₂O₉S Molecular Weight: 605,28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001615 CAS-Nummer: 1667-99-8 |
Spezifikation Testergebnisse
Appearance | Dark brown to black Crystalline powder |
Infrared spectrum | Conforms |
UV spectrum | Complies |
UV Lambda max | 424 to 436 nm (c=0.03 g/l in H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...