Ponceau Xylidin, rein

Lieferant: Thermo Scientific
Achtung

Synonyme: CI 16150

190260250 190261000
ACRO190260250EA 81.8 EUR
ACRO190260250 ACRO190261000
Ponceau Xylidin, rein
Ponceau Xylidin

CAS No.: 3761-53-3

Formel: C₁₈H₁₆N₂O₇S₂
Molecular Weight: 480,42 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00003897
CAS-Nummer: 3761-53-3

Bestellen

Spezifikation Testergebnisse

Appearance Dark red to bordeaux Powder
Infrared spectrum Conforms
UV spectrum Complies

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR