Tartrazine 89%, rein

Lieferant: Thermo Scientific
Gefahr

Synonyme: FD and C Yellow No. 5, Filter Yellow, Acid Yellow 23, Sodium 5-oxo-1-(4-sulphonatophenyl)-4-((4-sulphonatophenyl)diazenyl)-4,5-dihydro-1H-pyrazole-3-carboxylate

191891000 191895000
ACRO191891000EA 45.3 EUR
ACRO191891000 ACRO191895000
Tartrazine 89%, rein
Tartrazine

CAS No.: 1934-21-0

Formel: C₁₆H₁₂N₄O₉S₂
Molecular Weight: 534,36 g/mol
Schmelzpunkt: >300 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00148908
CAS-Nummer: 1934-21-0

Bestellen

Spezifikation Testergebnisse

Appearance Orange Powder
Infrared spectrum Conforms
Dye content ≥85 %
Arsenic (As) ≤3 ppm
Cadmium (Cd) ≤1 ppm
Lead (Pb) ≤10 ppm
Mercury (Hg) ≤1 ppm
Heavy metals (as Pb) ≤40 ppm
Insoluble matter ≤0.2 % (in water)
Volatile matter ≤15 % chlorides and sulfates (calculated as sodium salts)
Subsidiary dyes ≤1 %
Dye intermediates ≤0.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR