Tetracarbonyldi-µ-chlorodirhodium(I) 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Di-μ-chloro-tetracarbonyldirhodium(I), Chlorodicarbonylrhodium(I) dimer

197600010 197602500
ACRO197600010EA 687 EUR
ACRO197600010 ACRO197602500
Tetracarbonyldi-µ-chlorodirhodium(I) 97%
Tetracarbonyldi-µ-chlorodirhodium(I)

Appearance: Red to brown Crystalline powder or crystals or needles

Formel: C₄Cl₂O₄Rh₂
Molecular Weight: 388.76 g/mol
Schmelzpunkt: 121 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00135610
CAS-Nummer: 14523-22-9

Bestellen

Spezifikation Testergebnisse

Appearance Red to brown Crystalline powder or crystals or needles
Infrared spectrum Conforms
Assay ≥96.0 % (ex Rh)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR