N,N-Dimethylmethyleneammonium iodide 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: N,N-Dimethylmethyleneiminium Iodide, Eschenmoser's Salt

201010100 201010500
ACRO201010100EA 97.7 EUR
ACRO201010100 ACRO201010500
N,N-Dimethylmethyleneammonium iodide 97%
N,N-Dimethylmethyleneammonium iodide

Appearance: Cream to yellow Crystalline powder

Formel: C₃H₈IN
Molecular Weight: 185,01 g/mol
Schmelzpunkt: 219 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00011810
CAS-Nummer: 33797-51-2

Bestellen

Spezifikation Testergebnisse

Appearance Cream to yellow Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR