Amberlite® IRA-900 (Cl) Ionenaustauscherharz

Lieferant: Thermo Scientific

Synonyme: Amberlite®, IRA-900 chloride form, Amberlite® IRA-900

202315000 202310025 202311000
ACRO202315000EA 84.4 EUR
ACRO202315000 ACRO202310025 ACRO202311000
Amberlite® IRA-900 (Cl) Ionenaustauscherharz
Amberlite® IRA-900 (Cl)

CAS No.: 9050-97-9

Lagertemperatur: Raumtemperatur
MDL: MFCD00132712
CAS-Nummer: 9050-97-9

Bestellen

Spezifikation Testergebnisse

Appearance Cream to beige to light brown Beads
Particle size ≤3.0 % (> 1.180 mm)
Harmonic mean size 0.650 to 0.820 mm
Uniformity coefficient ≤1.8
Moisture holding capacity 58 to 64 % (Cl-)
Total volume capacity ≥1.0 eq/l (Cl-)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR