Vinblastine sulphate 96,0-102,0%

Lieferant: Thermo Scientific
Gefahr

203450050 203450250
ACRO203450050EA 90.7 EUR
ACRO203450050 ACRO203450250
Vinblastine sulphate 96,0-102,0%
Vinblastine sulphate

Appearance: White to light yellow Crystalline powder

Formel: C₄₆H₆₀N₄O₁₃S
Schmelzpunkt: 267 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00082457
CAS-Nummer: 143-67-9

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Water ≤15 %
HPLC 96.0 to 102.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR