(-)-Verbenon 94%

Lieferant: Thermo Scientific
Achtung

Synonyme: (-)-2-Pinen-4-one, (1S,5S)-4,6,6-Trimethylbicyclo[3.1.1]hept-3-en-2-one, (-)-4,6,6-Trimethylbicyclo[3.1.1]hept-3-en-2-one, (1S)-(-)-Verbenone

203630100 203630500
ACRO203630100EA 46.3 EUR
ACRO203630100 ACRO203630500
(-)-Verbenon 94%
(-)-Verbenone

Appearance: Clear colorless to yellow Liquid

Formel: C₁₀H₁₄O
Molecular Weight: 150,22 g/mol
Siedepunkt: 227…228 °C (760 mmHg)
Dichte: 0,97 g/cm³
Flammpunkt: 85 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001343
CAS-Nummer: 1196-01-6

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4950 to 1.4970 (20°C, 589 nm)
GC ≥92 %
Specific gravity (25°C) 0.96 to 0.98
Specific optical rotation -150° to -250° (20°C, 589 nm) (C=4, CHCl3) ,reported only for information

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR