Hydroxy naphthol blue disodium salt ACS
Lieferant: Thermo Scientific
Achtung
Synonyme: Hydroxynaphthol blue sodium salt, Hydroxy napthol blue, Sodium (E)-3-hydroxy-4-((2-hydroxy-6-sulpho-3-sulphonatonaphthalen-1-yl)diazenyl)naphthalene-1-sulphonate
204880050 204881000 204885000
ACRO204880050EA 25.8 EUR
ACRO204880050 ACRO204881000 ACRO204885000
Hydroxy naphthol blue disodium salt ACS
Hydroxy naphthol blue disodium salt
CAS No.: 165660-27-5
Formel: C₂₀H₁₂N₂O₁₁S₃·2Na
Molecular Weight: 598,479 g/mol
Schmelzpunkt: 274 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004075
CAS-Nummer: 165660-27-5
Bestellen
Spezifikation Testergebnisse
Appearance Blue to dark purple Crystalline powder or crystals
Infrared spectrum Conforms
Suitability (for Ca determination) Passes test