Ethylenedioxybis(trimethylsilane) 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1,2-Bis(trimethylsilyloxy)ethane, Ethylene Glycol Bis(trimethylsilyl Ether), 1,2-Bis(trimethylsiloxy)ethane
209580100
209580500
ACRO209580100EA
42.5
EUR
ACRO209580100
ACRO209580500
Ethylenedioxybis(trimethylsilane) 98%
Ethylenedioxybis(trimethylsilane)
Appearance: Clear colorless to light yellow Liquid
Formel:
C₈H₂₂O₂Si₂ Molecular Weight: 206,43 g/mol Siedepunkt: 165…166 °C (760 mmHg) Dichte: 0,84 g/cm³ Flammpunkt: 46 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009640 CAS-Nummer: 7381-30-8 UN: 1993 ADR: 3,III |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4020 to 1.4040 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...