Lawesson-Reagenz 99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2,4-Bis(4-methoxyphenyl)-2,4-dithioxo-1,3,2,4- dithiadiphosphetane, 2,4-Bis(4-methoxyphenyl)-1,3-dithia-2,4-diphosphetane 2,4-Disulphide

210890250 210891000 210895000
ACRO210890250EA 45.8 EUR
ACRO210890250 ACRO210891000 ACRO210895000
Lawesson-Reagenz 99%
Lawesson's reagent

CAS No.: 19172-47-5

Formel: C₁₄H₁₄O₂P₂S₄
Molecular Weight: 404,45 g/mol
Schmelzpunkt: 228…231 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005171
CAS-Nummer: 19172-47-5
UN: 3134
ADR: 4.3,II

Bestellen

Spezifikation Testergebnisse

Appearance Light yellow to beige Powder
Infrared spectrum Conforms
Melting point 220 to 226 °C
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR