Potassium hydrogen monopersulphate >4,5% (aktiver Sauerstoff), reinst
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
KMPS, Potassium peroxomonosulphate, Oxone, PMPS, Potassium monopersulphate, Potassium monopersulphate triple salt
211360010
211361000
211360025
ACRO211360010EA
74.6
EUR
ACRO211360010
ACRO211361000
ACRO211360025
Potassium hydrogen monopersulphate >4,5% (aktiver Sauerstoff), reinst
Potassium hydrogen monopersulphate
CAS No.: 37222-66-5
Formel:
H₃K₅O₁₈S₄ Molecular Weight: 614.76 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040551 CAS-Nummer: 37222-66-5 UN: 3260 ADR: 8 II |
Spezifikation Testergebnisse
Appearance | White Crystalline or granular powder |
Water | ≤0.5 % |
Titration | ≥4.5 % (Active oxygen) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...