Stains-all 97%, rein

Lieferant: Thermo Scientific
Achtung

Synonyme: 1-Ethyl-2-[3-(1-ethylnaphtho[1,2-d]thiazolin-2-ylidene)-2-methylpropenyl]naphtho[1,2-d]thiazolium bromide, 3,3′-Diethyl-9-methyl-4,5,4′,5′-dibenzothiacarbocyanine

213510010 213510050 213512500
ACRO213510010EA 97.9 EUR
ACRO213510010 ACRO213510050 ACRO213512500
Stains-all 97%, rein
Stains-all

CAS No.: 7423-31-6

Formel: C₃₀H₂₇BrN₂S₂
Molecular Weight: 559,584 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00078788
CAS-Nummer: 7423-31-6

Bestellen

Spezifikation Testergebnisse

Appearance Dark grey to dark purple Powder
Infrared spectrum Conforms
UV spectrum Complies
TLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR