4-Amino-2-mercaptopyrimidin 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Amino-2-mercaptopyrimidine, 4-Aminopyrimidine-2-thiol, 4-Amino-2-sulphanylpyrimidine

214770010 214770050
ACRO214770010EA 49.7 EUR
ACRO214770010 ACRO214770050
4-Amino-2-mercaptopyrimidin 97%
4-Amino-2-mercaptopyrimidine
Formel: C₄H₅N₃S
Molecular Weight: 127.1676 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00006075
CAS-Nummer: 333-49-3

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to brown Powder
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR