Chlorhexidine diacetate hydrate 98%
Lieferant: Thermo Scientific
|
Gefahr
|
214980050
214980250
ACRO214980050EA
53.1
EUR
ACRO214980050
ACRO214980250
Chlorhexidine diacetate hydrate 98%
Chlorhexidine diacetate hydrate
Appearance: White to off-white Powder
Formel:
C₂₂H₃₀Cl₂N₁₀·2C₂H₄O₂·xH₂O Schmelzpunkt: 153…157 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150042 CAS-Nummer: 206986-79-0 UN: 3077 ADR: 9,III |
Spezifikation Testergebnisse
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤3.5 % (105°C) |
Titration with HClO4 | ≥97.5 % (On dry substance) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...