N, O-Bis (Trimethylsilyl)Hydroxylamin 97%

Lieferant: Thermo Scientific
Gefahr

215980050 215980010 215980250
ACRO215980050EA 117 EUR
ACRO215980050 ACRO215980010 ACRO215980250
N, O-Bis (Trimethylsilyl)Hydroxylamin 97%
N, O-Bis (Trimethylsilyl)Hydroxylamin

Appearance: Clear colorless Liquid

Formel: C₆H₁₉NOSi₂
Molecular Weight: 177,39 g/mol
Siedepunkt: 78…80 °C (100 mmHg)
Dichte: 0,83 g/cm³
Flammpunkt: 28 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00008266
CAS-Nummer: 22737-37-7
UN: 2920
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR