Poly(oxy(methylsilylene))

Lieferant: Thermo Scientific

ACRO175095000EA 124 EUR
ACRO175095000 ACRO175091000
Poly(oxy(methylsilylene))
Poly(oxy(methylsilylene))

Appearance: Clear colorless Viscous liquid

Formel: C₅H₁₁OSi
Dichte: 1 g/cm³
Flammpunkt: 121 °C
Lagertemperatur: Raumtemperatur
CAS-Nummer: 9004-73-3

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless Viscous liquid
Infrared spectrum Conforms
Refractive index 1.3970 to 1.4010 (20°C, 589 nm)
Viscosity 15 to 40 mPa.s (at 20°C)
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR