methyl cyclobutanecarboxylate 98%

Lieferant: Thermo Scientific

Synonyme: Methyl cyclobutanecarboxylate

365540050 365540250
ACRO365540050EA 47 EUR
ACRO365540050 ACRO365540250
methyl cyclobutanecarboxylate 98%
methyl cyclobutanecarboxylate

CAS No.: 765-85-5

Formel: C₆H₁₀O₂
Molecular Weight: 114,144 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00095142
CAS-Nummer: 765-85-5

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4260 to 1.4300 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR