4-Brompiperidinhydrobromid 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
368310010
368310100
ACRO368310010EA
35.7
EUR
ACRO368310010
ACRO368310100
4-Brompiperidinhydrobromid 98%
4-Bromopiperidine hydrobromide
Appearance: White Crystalline powder
Formel:
C₅H₁₁Br₂N Molecular Weight: 244.9600 g/mol Schmelzpunkt: 199…202 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191858 CAS-Nummer: 54288-70-9 |
Spezifikation Testergebnisse
Appearance | White to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 191 to 196 °C |
Titration Argentometric | 97.5 to 102.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...