Pyrrole-2-carbonitrile 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
368350010
368350050
ACRO368350010EA
46.8
EUR
ACRO368350010
ACRO368350050
Pyrrole-2-carbonitrile 97%
Pyrrole-2-carbonitrile
Appearance: Clear yellow to orange Liquid
Formel:
C₅H₄N₂ Molecular Weight: 92.101 g/mol Siedepunkt: 115…120 °C (10 mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234061 CAS-Nummer: 4513-94-4 UN: 3276 ADR: 6.1 III |
Spezifikation Testergebnisse
Appearance | Clear yellow to orange Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5480 to 1.5510 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...