1-Dimethylamino-2-propyne 97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: N,N-Dimethylpropargylamine, N,N-Dimethyl-2-propyn-1-amine

162250050 162250250
ACRO162250050EA 36.5 EUR
ACRO162250050 ACRO162250250
1-Dimethylamino-2-propyne 97%
1-Dimethylamino-2-propyne

Appearance: Clear colorless to yellow Liquid

Formel: C₅H₉N
Molecular Weight: 83,13 g/mol
Siedepunkt: 80…81 °C (760 mmHg)
Dichte: 0,772 g/cm³
Flammpunkt: –7 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00008575
CAS-Nummer: 7223-38-3
UN: 2733
ADR: 3,II

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4190 to 1.4220 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR