Valinomycin 98%

Lieferant: Thermo Fisher Scientific
Gefahr

385120100 385120500
ACRO385120100EA 84.8 EUR
ACRO385120100 ACRO385120500
Valinomycin 98%
Valinomycin

Appearance: White Crystalline powder

Formel: C₅₄H₉₀O₁₈N₆
Schmelzpunkt: 190 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00005114
CAS-Nummer: 2001-95-8
UN: 2811
ADR: 6.1,I

Bestellen

Spezifikation Testergebnisse

Appearance White Powder
Melting point 186 to 191°C
Solubility (50mg/ml CH2Cl2) Clear colorless solution
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR