O-Benzyl-N-(tert-butoxycarbonyl)-D-serine 98%

Lieferant: Thermo Fisher Scientific

Synonyme: Boc-D-Ser(Bzl)-OH, N-Boc-O-benzyl-D-serine

334670050 334670250
ACRO334670050EA 156 EUR
ACRO334670050 ACRO334670250
O-Benzyl-N-(tert-butoxycarbonyl)-D-serine 98%
O-Benzyl-N-(tert-butoxycarbonyl)-D-serine

Appearance: White Powder

Formel: C₁₅H₂₁NO₅
Molecular Weight: 295,34 g/mol
Schmelzpunkt: 58…61 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00038248
CAS-Nummer: 47173-80-8

Bestellen

Spezifikation Testergebnisse

Appearance White Powder
Infrared spectrum Conforms
Melting point 58 to 63 °C
HPLC ≥97.5 %
Specific optical rotation -20° to -24° (20 °C, 589 nm) (c=1, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR