tert-Butyl (3R)-3-aminobutanoate 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
(R)-tert-Butyl 3-aminobutanoate
337955000
337950025
ACRO337955000EA
154
EUR
ACRO337955000
ACRO337950025
tert-Butyl (3R)-3-aminobutanoate 95%
tert-Butyl (3R)-3-aminobutanoate
CAS No.: 158849-23-1
Formel:
C₈H₁₇NO₂ Molecular Weight: 159.229 g/mol Siedepunkt: 50 °C (.7 mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00798293 CAS-Nummer: 158849-23-1 UN: 2735 ADR: 8 III |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4240 to 1.4260 (20°C, 589 nm) |
GC | ≥94 % |
Specific optical rotation | -21° to -27° (20°C, 589 nm) (C=1, CHCl3) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...