8-hydroxyquinoline-2-carbonitrile 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
8-Hydroxyquinoline-2-carbonitrile
347640010
347640050
ACRO347640010EA
47.9
EUR
ACRO347640010
ACRO347640050
8-hydroxyquinoline-2-carbonitrile 98%
8-hydroxyquinoline-2-carbonitrile
Appearance: Yellow to brown Crystalline powder
Formel:
C₁₀H₆N₂O Molecular Weight: 170.171 g/mol Schmelzpunkt: 131…136 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00216717 CAS-Nummer: 6759-78-0 |
Spezifikation Testergebnisse
Appearance | Yellow to brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 131 to 136 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...