Benzyl(triphenylphosphoranylidene)acetate 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Benzyl 2-(triphenylphosphoranylidene)acetate

377220050 377220250 377221000
ACRO377220050EA 60.9 EUR
ACRO377220050 ACRO377220250 ACRO377221000
Benzyl(triphenylphosphoranylidene)acetate 97%
Benzyl(triphenylphosphoranylidene)acetate

CAS No.: 15097-38-8

Formel: C₂₇H₂₃O₂P
Molecular Weight: 410,453 g/mol
Schmelzpunkt: 120…127 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00191787
CAS-Nummer: 15097-38-8
UN: 3464
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 120 to 127 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR