Pivalic acid hydrazide 98+%

Lieferant: Thermo Scientific
Achtung

Synonyme: Pivalohydrazide, Pivaloyl hydrazide

377750010 377750050
ACRO377750010EA 37 EUR
ACRO377750010 ACRO377750050
Pivalic acid hydrazide 98+%
Pivalic acid hydrazide

Appearance: White Crystals

Formel: C₅H₁₂N₂O
Molecular Weight: 116,164 g/mol
Siedepunkt: 95…110 °C (1mbar)
Schmelzpunkt: 65…67 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01310839
CAS-Nummer: 42826-42-6
UN: 1325
ADR: 4.1,II

Bestellen

Spezifikation Testergebnisse

Appearance White Crystals
Infrared spectrum Conforms
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR