Phenethyl isocyanate 99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Isocyanic Acid Phenethyl Ester, (2-Isocyanatoethyl)benzene, Isocyanic Acid 2-Phenylethyl Ester, 2-Phenylethyl Isocyanate
380660050
380660250
ACRO380660050EA
95
EUR
ACRO380660050
ACRO380660250
Phenethyl isocyanate 99%
Phenethyl isocyanate
Appearance: Colorless to yellow Liquid
Formel:
C₉H₉NO Molecular Weight: 147,18 g/mol Siedepunkt: 210 °C (760 mmHg) Dichte: 1,063 g/cm³ Flammpunkt: 99 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00037044 CAS-Nummer: 1943-82-4 UN: 2927 ADR: 6.1,II |
Spezifikation Testergebnisse
Appearance | Colorless to yellow Liquid (May develop some turbidity or precipitate) |
Infrared spectrum | Conforms |
Refractive index | 1.5210 to 1.5230 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...