4-Borono-L-phenylalanine 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
H-Phe(4-B)-OH, L-4-(2-Amino-2-carboxyethyl)benzeneboronic acid, (S)-2-Amino-3-(4-boronophenyl)propanoic acid
358931000
358932500
ACRO358931000EA
77.6
EUR
ACRO358931000
ACRO358932500
4-Borono-L-phenylalanine 97%
4-Borono-L-phenylalanine
CAS No.: 76410-58-7
Formel:
C₉H₁₂BNO₄ Schmelzpunkt: 283…293 °C (Zers.) Lagertemperatur: Kühlschrank |
MDL:
MFCD01075172 CAS-Nummer: 76410-58-7 |
Spezifikation Testergebnisse
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...