N,N-Dimethylanilinium tetrakis(pentafluorophenyl)borate 98%
Lieferant: Thermo Fisher Scientific
Gefahr
Synonyme: Dimethylanilinium tetrakis (pentafluorophenyl)borate, N,N-Dimethylanilinium tetra(pentafluorophenyl)borate, N,N-Dimethylbenzenaminium tetrakis(perfluorophenyl)borate
382400010 382400050
ACRO382400010EA 232 EUR
ACRO382400010 ACRO382400050
N,N-Dimethylanilinium tetrakis(pentafluorophenyl)borate 98%
N,N-Dimethylanilinium tetrakis(pentafluorophenyl)borate
Appearance: White to off-white Powder
Formel: C₃₂H₁₂BF₂₀N
Molecular Weight: 801.23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12913898
CAS-Nummer: 118612-00-3
Bestellen
Spezifikation Testergebnisse
Appearance White to off-white Powder (May have a blue hue)
Infrared spectrum Conforms
NMR (19F-NMR) ≥97.5 %