(1S,3R)-N-Boc-1-aminocyclopentane-3-carboxylic acid 95%, 98% ee

Lieferant: Thermo Fisher Scientific

Synonyme: (1R,3S)-(+)-3-Aminocyclopentane-1-carboxylic acid, N-BOC protected, (1R,3S)-3-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid

361840010 361842500
ACRO361840010EA 519 EUR
ACRO361840010 ACRO361842500
(1S,3R)-N-Boc-1-aminocyclopentane-3-carboxylic acid 95%, 98% ee
(1S,3R)-N-Boc-1-aminocyclopentane-3-carboxylic acid

Appearance: White Powder

Formel: C₁₁H₁₉NO₄
Molecular Weight: 229.276 g/mol
Schmelzpunkt: 96.1 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01320858
CAS-Nummer: 161660-94-2

Bestellen

Spezifikation Testergebnisse

Appearance White Powder
Infrared spectrum Conforms
HPLC ≥94 %
Enantiomeric excess ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR