4-Fluoro-L-2-phenylglycine ≥95%, ee 98%

Lieferant: Thermo Fisher Scientific

Synonyme: 4-Fluoro-L-phenylglycine, (S)-2-Amino-2-(4-fluorophenyl)acetic Acid, H-Phg(4-F)-OH, (S)-4-Fluorophenylglycine

362050010 362050050
ACRO362050010EA 198 EUR
ACRO362050010 ACRO362050050
4-Fluoro-L-2-phenylglycine ≥95%, ee 98%
4-Fluoro-L-2-phenylglycine

Appearance: Off-white Powder

Formel: C₈H₈FNO₂
Molecular Weight: 169.16 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00213799
CAS-Nummer: 19883-57-9

Bestellen

Spezifikation Testergebnisse

Appearance Off-white Powder
Infrared spectrum Conforms
HPLC ≥94 %
Enantiomeric excess ≥97.5 % (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR