(R)-N-Boc-allylglycine 95%, 98% ee
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
N-Boc-allylglycine, (2R)-2-{[(tert-butoxy)carbonyl]amino}pent-4-enoate, (R)-2-((tert-Butoxycarbonyl)amino)pent-4-enoic acid
362840010
362845000
ACRO362840010EA
158
EUR
ACRO362840010
ACRO362845000
(R)-N-Boc-allylglycine 95%, 98% ee
(R)-N-Boc-allylglycine
Appearance: Clear light yellow Viscous liquid
Formel:
C₁₀H₁₆NO₄ Molecular Weight: 215.25 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD01320882 CAS-Nummer: 170899-08-8 |
Spezifikation Testergebnisse
Appearance | Clear light yellow Viscous liquid or paste |
Infrared spectrum | Conforms |
GC | ≥94 % |
Enantiomeric excess | ≥97.5 % |
Specific optical rotation | -9.5° to -12.5° (20°C, 589 nm) (c=1, methanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...