Celestine blue, rein

Lieferant: Thermo Scientific

ACRO195560500EA 320 EUR
ACRO195560500 ACRO195560100
Celestine blue, rein
Celestine blue
Formel: C₁₇H₁₈ClN₃O₄
Molecular Weight: 363,798 g/mol
Schmelzpunkt: 227…230 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011927
CAS-Nummer: 1562-90-9

Bestellen

Spezifikation Testergebnisse

Appearance Dark grey to dark brown to black Powder
Infrared spectrum Conforms
UV spectrum Complies
UV Lambda max 638 to 648 nm
Specific extinction E (1%/1cm) ≥700 (in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR