S-Methyl methanethiosulphonate (MMTS) 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Methanethiosulphonic Acid S-Methyl Ester, Methyl methanethiosulphonate

197390010 197390050 197390250 197391000
ACRO197390010EA 47.9 EUR
ACRO197390010 ACRO197390050 ACRO197390250 ACRO197391000
S-Methyl methanethiosulphonate (MMTS) 97%
S-Methyl methanethiosulphonate (MMTS)

Appearance: Clear yellow to orange Liquid

Formel: C₂H₆O₂S₂
Molecular Weight: 126,19 g/mol
Siedepunkt: 69…71 °C (,4 mmHg)
Dichte: 1,227 g/cm³
Flammpunkt: 87 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007565
CAS-Nummer: 2949-92-0
UN: 3334
ADR: 9,III

Bestellen

Spezifikation Testergebnisse

Appearance Clear yellow to orange Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR